accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOO

CAS RN 7472-43-7

IUPAC Name: 7-oxo-7-phenylheptanoic acid
Synonyms: 7-oxo-7-phenylheptanoic acid
Molecular Weight (g/mol): 220.27
Molecular Formula: C13H16O3
InChi Key: DOQWHEUDAHLEPT-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results