Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Benzoylhexanoic acid, 94%, Thermo Scientific Chemicals

Catalog No. AAL0843103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0843103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0843103 Supplier Thermo Scientific Chemicals Supplier No. L0843103
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7472-43-7 | Molecular Formula: C13H16O3 | Molecular Weight: 220.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7472-43-7
Molecular Formula C13H16O3
Molecular Weight (g/mol) 220.27
MDL Number MFCD00039608
InChI Key DOQWHEUDAHLEPT-UHFFFAOYSA-N
Synonym 6-benzoylhexanoic acid, 7-oxo-7-phenyl-heptanoic acid, 6-benzoyl hexanoic acid, benzeneheptanoic acid, z-oxo, 7-oxo-7-phenylheptanoicacid, #, benzeneheptanoic acid,, ae-oxo, alpha-oxo benzeneheptanoic acid, benzeneheptanoic acid,alpha-oxo
PubChem CID 344830
IUPAC Name 7-oxo-7-phenylheptanoic acid
SMILES OC(=O)CCCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 83°C to 86°C
Quantity 1 g
Beilstein 2694641
Formula Weight 220.27
Percent Purity 94%
Chemical Name or Material 6-Benzoylhexanoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.