accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHNO

CAS RN 7472-54-0

IUPAC Name: N-(4-methoxyphenyl)benzamide
Synonyms: N-(4-methoxyphenyl)benzamide
Molecular Weight (g/mol): 227.26
Molecular Formula: C14H13NO2
InChi Key: KEEBHMMBUBEEOV-UHFFFAOYSA-N
SMILES: COC1=CC=C(NC(=O)C2=CC=CC=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results