Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Benzanisidide 98.0+%, TCI America™
SDP

Catalog No. B081525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B081525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B081525G Supplier TCI America Supplier No. B081525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7472-54-0
Molecular Formula C14H13NO2
Molecular Weight (g/mol) 227.26
MDL Number MFCD00025788
InChI Key KEEBHMMBUBEEOV-UHFFFAOYSA-N
Synonym N-Benzoyl-p-anisidine, 4′C-Methoxybenzanilide, N-(4-Methoxyphenyl)benzamide
PubChem CID 139031
IUPAC Name N-(4-methoxyphenyl)benzamide
SMILES COC1=CC=C(NC(=O)C2=CC=CC=C2)C=C1

Specifications

Melting Point 155°C
Quantity 25 g
Formula Weight 227.26
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material p-Benzanisidide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.