accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3COHO

CAS RN 7473-98-5

IUPAC Name: 2-hydroxy-2-methyl-1-phenylpropan-1-one
Synonyms: benzoyl isopropanol
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: XMLYCEVDHLAQEL-UHFFFAOYSA-N
SMILES: CC(C)(O)C(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)