Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Hydroxy-2-methylpropiophenone

Catalog No. 40565550ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-565-550ML 50 mL
40-565-5250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-565-550ML Supplier Sigma Aldrich Supplier No. 40565550ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Hydroxy-2-methylpropiophenone is a radical photoinitiator (PI) molecule that can be used in the crosslinking of polymers by the exposure of UV radiation. It can be used in the development of UV curable resins for exterior coating applications.

Specifications

CAS 7473-98-5
Density 1.077 g/mL (at 25°C)
Boiling Point 102°C to 103°C (4 mmHg)
Molecular Formula C10H12O2
Refractive Index n20/D 1.533 (literature)
Linear Formula C6H5COC(CH3)2 OH
Quantity 50 mL
MDL Number MFCD00040710
Synonym 1-Hydroxy-1-methylethyl phenyl ketone; 1-Phenyl-2-methyl-2-hydroxypropanone; 2-Hydroxy-2-methyl-1-phenyl-1-propanone; 2-Methyl-3-phenyl-3-oxopropan-2-ol
Molecular Weight (g/mol) 164.2
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.