accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSH2N

CAS RN 747411-11-6

IUPAC Name: 3-ethoxybenzene-1-carbothioamide
Synonyms: 3-ethoxybenzenecarbothioamide
Molecular Weight (g/mol): 181.25
Molecular Formula: C9H11NOS
InChi Key: LXXDVDKIORLFFH-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC(=C1)C(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results