accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNONNOCH3

CAS RN 7477-67-0

IUPAC Name: 5,5'-dimethyl-2,2'-diphenyl-2H,2'H,3H,3'H,4H,4'H-[4,4'-bipyrazole]-3,3'-dione
Synonyms: 5,5'-dimethyl-2,2'-diphenyl-4H,4'H-[4,4'-bipyrazole]-3,3'-dione
Molecular Weight (g/mol): 346.39
Molecular Formula: C20H18N4O2
InChi Key: FORCWSNQDMPPOC-UHFFFAOYNA-N
SMILES: CC1=NN(C(=O)C1C1C(C)=NN(C1=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)