accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSOHNO

CAS RN 74772-77-3

IUPAC Name: 5-({4-[(1-methylcyclohexyl)methoxy]phenyl}methyl)-1,3-thiazolidine-2,4-dione
Synonyms: ciglitazone ciglitazona 环格列酮 ciglitazonum
Molecular Weight (g/mol): 333.45
Molecular Formula: C18H23NO3S
InChi Key: YZFWTZACSRHJQD-UHFFFAOYNA-N
SMILES: CC1(COC2=CC=C(CC3SC(=O)NC3=O)C=C2)CCCCC1
Quantity 
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results