Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Ciglitazone

Catalog No. 2309505MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-095-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-095-05MG Supplier MilliporeSigma™ Supplier No. 2309505MG
Add to Cart
Add to Cart

A potent thiazolinedione (TDZ) type anti-hyperglycemic agent and a selective PPARγ agonist (EC50 = 3 μM).

A potent thiazolinedione (TDZ) type anti-hyperglycemic agent and a selective PPARγ agonist (EC50 = 3μM).
TRUSTED_SUSTAINABILITY

Specifications

Color Tan
Description 98% by TLC
Target Proteins Selective PPARγ agonist
CAS 74772-77-3
Molecular Formula C18H23NO3S
Formula Weight 333.5g/mol
Solubility DMSO or EtOH
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.