accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHOSHO

CAS RN 74772-78-4

IUPAC Name: 5-[(4-hydroxyphenyl)methyl]-1,3-thiazolidine-2,4-dione
Synonyms: 5-[(4-hydroxyphenyl)methyl]-1,3-thiazolidine-2,4-dione
Molecular Weight (g/mol): 223.25
Molecular Formula: C10H9NO3S
InChi Key: NKOHRVBBQISBSB-UHFFFAOYNA-N
SMILES: OC1=CC=C(CC2SC(=O)NC2=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results