accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7481-90-5

IUPAC Name: 1-{2,6-dioxabicyclo[3.2.0]heptan-3-yl}-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 1-{2,6-dioxabicyclo[3.2.0]heptan-3-yl}-5-methyl-3H-pyrimidine-2,4-dione
Molecular Weight (g/mol): 224.22
Molecular Formula: C10H12N2O4
InChi Key: OAWLMYIJZBBZTP-UHFFFAOYNA-N
SMILES: CC1=CN(C2CC3OCC3O2)C(=O)NC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results