Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3,5-Anhydro-2-deoxy-beta-D-threo-pentofuranosyl)thymine 98.0+%, TCI America™
SDP

Catalog No. A207325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A207325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A207325G Supplier TCI America Supplier No. A207325G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7481-90-5
Molecular Formula C10H12N2O4
Molecular Weight (g/mol) 224.216
InChI Key OAWLMYIJZBBZTP-XLPZGREQSA-N
Synonym 3′C,5′C-Anhydrothymidine
PubChem CID 14069470
IUPAC Name 1-[(1S,3R,5R)-4,7-dioxabicyclo[3.2.0]heptan-3-yl]-5-methylpyrimidine-2,4-dione
SMILES CC1=CN(C(=O)NC1=O)C2CC3C(O2)CO3

Specifications

Melting Point 196°C
Color White
Quantity 25 g
Formula Weight 224.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(3,5-Anhydro-2-deoxy-beta-D-threo-pentofuranosyl)thymine
EINECSNumber (5)-6691
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.