accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 74815-81-9

IUPAC Name: 6,7-dibromonaphthalene-2,3-dicarbonitrile
Synonyms: 6,7-dibromonaphthalene-2,3-dicarbonitrile
Molecular Weight (g/mol): 335.99
Molecular Formula: C12H4Br2N2
InChi Key: YHUVAAVMNCSZQN-UHFFFAOYSA-N
SMILES: BrC1=CC2=CC(C#N)=C(C=C2C=C1Br)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results