Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dibromo-6,7-dicyanonaphthalene 98.0+%, TCI America™
SDP

Catalog No. D22821G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D22821G Supplier TCI America Supplier No. D22821G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 74815-81-9
Molecular Formula C12H4Br2N2
Molecular Weight (g/mol) 335.986
MDL Number MFCD00191423
InChI Key YHUVAAVMNCSZQN-UHFFFAOYSA-N
Synonym 6,7-Dibromonaphthalene-2,3-dicarbonitrile
PubChem CID 630682
IUPAC Name 6,7-dibromonaphthalene-2,3-dicarbonitrile
SMILES C1=C2C=C(C(=CC2=CC(=C1Br)Br)C#N)C#N

Specifications

Color Yellow
Quantity 1 g
UN Number 3439
Formula Weight 335.99
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dibromo-6,7-dicyanonaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.