accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 74853-62-6

HOONNHN

CAS RN 74853-62-6

IUPAC Name: 3-[1-(triphenylmethyl)-1H-imidazol-4-yl]-2-[(triphenylmethyl)amino]propanoic acid
Synonyms: 2-[(triphenylmethyl)amino]-3-[1-(triphenylmethyl)imidazol-4-yl]propanoic acid
Molecular Weight (g/mol): 639.8
Molecular Formula: C44H37N3O2
InChi Key: FFVVHORODDZQPF-UHFFFAOYSA-N
SMILES: OC(=O)C(CC1=CN(C=N1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)