accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 749188-33-8

IUPAC Name: N'-hydroxy-4-(piperazin-1-yl)benzene-1-carboximidamide
Synonyms: N'-hydroxy-4-(piperazin-1-yl)benzenecarboximidamide
Molecular Weight (g/mol): 220.28
Molecular Formula: C11H16N4O
InChi Key: IWRIWCGOFIEEIB-UHFFFAOYSA-N
SMILES: NC(=NO)C1=CC=C(C=C1)N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results