accessibility menu, dialog, popup

UserName

Viewing profile as user:
CaCH3OOCH3OO(E)(E)(E)(E)

CAS RN 7492-55-9

IUPAC Name: calcium bis((2E,4E)-hexa-2,4-dienoate)
Synonyms: calcium sorbate
Molecular Weight (g/mol): 262.32
Molecular Formula: C12H14CaO4
InChi Key: MCFVRESNTICQSJ-RJNTXXOISA-L
SMILES: [Ca++].C\C=C\C=C\C([O-])=O.C\C=C\C=C\C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results