Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Sorbate 95.0+%, TCI America™
SDP

Catalog No. S005425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S005425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S005425G Supplier TCI America Supplier No. S005425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7492-55-9
Molecular Formula C12H14CaO4
Molecular Weight (g/mol) 262.32
MDL Number MFCD00043805
InChI Key MCFVRESNTICQSJ-RJNTXXOISA-L
Synonym Sorbic Acid Calcium Salt
PubChem CID 6433506
IUPAC Name calcium bis((2E,4E)-hexa-2,4-dienoate)
SMILES [Ca++].C\C=C\C=C\C([O-])=O.C\C=C\C=C\C([O-])=O

Specifications

Quantity 25 g
Formula Weight 262.32
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Calcium Sorbate
EINECSNumber (2)-1076
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.