accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7492-70-8

H3COOCH3OOCH3

CAS RN 7492-70-8

IUPAC Name: 1-butoxy-1-oxopropan-2-yl butanoate
Synonyms: butyl butyrolactate
Molecular Weight (g/mol): 216.28
Molecular Formula: C11H20O4
InChi Key: NORZZKKLCYMBBF-UHFFFAOYNA-N
SMILES: CCCCOC(=O)C(C)OC(=O)CCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)