Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl O-Butyryllactate 98.0+%, TCI America™
SDP

Catalog No. B434425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B434425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B434425ML Supplier TCI America Supplier No. B434425ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7492-70-8
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.277
MDL Number MFCD00027213
InChI Key NORZZKKLCYMBBF-UHFFFAOYSA-N
Synonym 2-(Butyryloxy)propionic Acid Butyl Ester, Butyl 2-(Butyryloxy)propionate, O-Butyryllactic Acid Butyl Ester
PubChem CID 24114
IUPAC Name (1-butoxy-1-oxopropan-2-yl) butanoate
SMILES CCCCOC(=O)C(C)OC(=O)CCC

Specifications

Color Colorless
Boiling Point 90°C
Quantity 25 mL
Formula Weight 216.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl O-Butyryllactate
EINECSNumber (2)-1396&(2)-2737
RTECSNumber ES8123000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.