accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COH3CNCS

CAS RN 749261-40-3

IUPAC Name: 1-(1-isothiocyanatoethyl)-4-methoxybenzene
Synonyms: 1-(1-isothiocyanatoethyl)-4-methoxybenzene
Molecular Weight (g/mol): 193.26
Molecular Formula: C10H11NOS
InChi Key: HOWJPQXDLZDTBF-UHFFFAOYNA-N
SMILES: COC1=CC=C(C=C1)C(C)N=C=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results