accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3HO

CAS RN 74938-11-7

IUPAC Name: 7-(dipropylamino)-5,6,7,8-tetrahydronaphthalen-2-ol
Synonyms: 7-OH-dpat
Molecular Weight (g/mol): 247.38
Molecular Formula: C16H25NO
InChi Key: BLYMJBIZMIGWFK-UHFFFAOYNA-N
SMILES: CCCN(CCC)C1CCC2=CC=C(O)C=C2C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results