Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-7-Hydroxy-2-di-n-propylaminotetralin hydrobromide, Thermo Scientific™

Catalog No. AAJ61065MF
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61065MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61065MF Supplier Thermo Scientific Chemicals Supplier No. J61065MF

missing translation for 'validMainframeMsg'

Selective agonist for D3 dopamine

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74938-11-7
Molecular Formula C16H26BrNO
Molecular Weight (g/mol) 328.294
MDL Number MFCD00153809
InChI Key ODNDMTWHRYECKX-UHFFFAOYSA-N
Synonym 7-dipropylamino-5,6,7,8-tetrahydronaphthalen-2-ol hydrobromide, 7-hydroxy-dpat hydrobromide, +/--7-oh-dpat, 7-oh-dpat hydrobromide, r +-7-hydroxy-dpat hydrobromide, +/--7-hydroxy-dpat hydrobromide, +/--7-hydroxy-2-di-n-propylamino tetralin hydrobromide, +/--7-hydroxy-2-dipropylaminotetralin hydrobromide, r +-7-hydroxy dipropylamino tetralin hydrobromide, y-7-hydroxy-dpat hydrobromide
PubChem CID 11957566
IUPAC Name 7-(dipropylamino)-5,6,7,8-tetrahydronaphthalen-2-ol;hydrobromide
SMILES CCCN(CCC)C1CCC2=C(C1)C=C(C=C2)O.Br

Specifications

Melting Point 168°C to 170°C
Color White
Quantity 50 mg
Formula Weight 329.29
Physical Form Powder
Chemical Name or Material (+/-)-7-Hydroxy-2-di-n-propylaminotetralin hydrobromide
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.