CAS RN 7496-46-0
IUPAC Name:
8-(bromomethyl)quinoline
Synonyms:
8-(bromomethyl)quinoline
Molecular Weight (g/mol):
222.09
Molecular Formula:
C10H8BrN
InChi Key:
IAAUGSYGHOFWEW-UHFFFAOYSA-N
SMILES:
BrCC1=C2N=CC=CC2=CC=C1
1
–
1
of
1
results
1
Sigma Aldrich 8-(Bromomethyl)quinoline
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.