accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFSOOOFFFSOOOFFFSOOOGa

CAS RN 74974-60-0

IUPAC Name: gallium(3+) tritrifluoromethanesulfonate
Synonyms: gallium(3+) tritriflate
Molecular Weight (g/mol): 516.91
Molecular Formula: C3F9GaO9S3
InChi Key: BVLXNPRUOXPBII-UHFFFAOYSA-K
SMILES: [Ga+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results