accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 74974-61-1

IUPAC Name: aluminium(3+) tritrifluoromethanesulfonate
Synonyms: aluminium(3+) tritriflate
Molecular Weight (g/mol): 474.17
Molecular Formula: C3AlF9O9S3
InChi Key: FKOASGGZYSYPBI-UHFFFAOYSA-K
SMILES: [Al+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results