accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NSOOOOONOO(E/Z)

CAS RN 749872-43-3

IUPAC Name: [(1,1-dioxo-1λ⁶-thiolan-3-yl)(methyl)carbamoyl]methyl 3-(3-nitrophenyl)prop-2-enoate
Synonyms: [(1,1-dioxo-1λ⁶-thiolan-3-yl)(methyl)carbamoyl]methyl 3-(3-nitrophenyl)prop-2-enoate
Molecular Weight (g/mol): 382.39
Molecular Formula: C16H18N2O7S
InChi Key: URVRJYLSUVXWBC-UHFFFAOYNA-N
SMILES: CN(C1CCS(=O)(=O)C1)C(=O)COC(=O)C=CC1=CC(=CC=C1)[N+]([O-])=O
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results