Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ KLF5 Inhibitor, CID 5951923

Catalog No. 42262510MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-262-510MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-262-510MG Supplier MilliporeSigma™ Supplier No. 42262510MG
Add to Cart
Add to Cart

The KLF5 Inhibitor, CID 5951923 controls the biological activity of KLF5.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 99% by HPLC
CAS 749872-43-3
Molecular Formula C16H18N2O7S
Formula Weight 382.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.