accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 75-21-8

IUPAC Name: oxirane
Synonyms: ethylene oxide anproline anprolene caswell no. 443 eto - ethylene oxide etilene (ossido di) ETO aethylenoxid merpol T-gas
Molecular Weight (g/mol): 44.05
Molecular Formula: C2H4O
InChi Key: IAYPIBMASNFSPL-UHFFFAOYSA-N
SMILES: C1CO1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
Solvent or Matrix 
  • (1)
Technique 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results