Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Beta-Cyclodextrin

Catalog No. 779105100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-910-5100G 100 mL
77-910-51KG 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 77-910-5100G Supplier Sigma Aldrich Supplier No. 779105100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Beta-Cyclodextrin

Specifications

CAS 75-21-8
Molecular Formula C2H4O
Linear Formula C2H4O
MDL Number MFCD00014482
Quantity 100 mL
Synonym Cavamax(R) W7; beta-Cyclodextrin; Caraway; Cycloheptaamylose; Cyclomaltoheptaose; Schardinger beta-Dextrin
Molecular Weight (g/mol) 44.05 g/mol
Biological Activity Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.