accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 75-27-4

IUPAC Name: bromodichloromethane
Synonyms: bromodichloromethane ion(1-) bromodichloromethane C-H-br-cl2 bromodichloromethane, ion(1-) bromodichloromethane (substance) ccris 819 BDCM
Molecular Weight (g/mol): 163.82
Molecular Formula: CHBrCl2
InChi Key: FMWLUWPQPKEARP-UHFFFAOYSA-N
SMILES: ClC(Cl)Br
Molecular Weight (g/mol) 
  • (2)
  • (3)
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)