Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromodichloromethane (stabilized with Ethanol) 96.0+%, TCI America™
SDP

Catalog No. B101125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B101125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B101125G Supplier TCI America Supplier No. B101125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 75-27-4
Molecular Formula CHBrCl2
Molecular Weight (g/mol) 163.823
MDL Number MFCD00000824
InChI Key FMWLUWPQPKEARP-UHFFFAOYSA-N
Synonym dichlorobromomethane, methane, bromodichloro, bromo dichloro methane, dichloromonobromomethane, monobromodichloromethane, bdcm, bromodichlormethane, unii-7ln464ch2o, ccris 819, stabilized with ethanol
PubChem CID 6359
ChEBI CHEBI:34591
IUPAC Name bromo(dichloro)methane
SMILES C(Cl)(Cl)Br

Specifications

Color Yellow
Boiling Point 87°C
Quantity 25 g
Formula Weight 163.82
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Bromodichloromethane (stabilized with Ethanol)
RTECSNumber PA5310000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.