accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3Cl

CAS RN 75-29-6

IUPAC Name: 2-chloropropane
Synonyms: 2-chloropropane
Molecular Weight (g/mol): 78.54
Molecular Formula: C3H7Cl
InChi Key: ULYZAYCEDJDHCC-UHFFFAOYSA-N
SMILES: CC(C)Cl
Percent Purity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)