Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloropropane, 99%, Thermo Scientific™

Catalog No. AAL04535AP
Change view
Click to view available options
Quantity:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL04535AP 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL04535AP Supplier Thermo Scientific Chemicals Supplier No. L04535AP

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Chloropropane is used as a solvent and an intermediate. It shows anesthetic properties. Further, it is used as an alternative foam-blowing agent. It is used to prepare chlorodiethylisopropylsilane.

Solubility
Miscible with water, ethanol, methanol, benzene and ether.

Notes
Incompatible with strong oxidizing agents, strong bases and metals.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-29-6
Boiling Point 35°C to 36°C
Flash Point −35°C (−31°F)
Odor Slightly Sweetish
Molecular Formula C3H7Cl
Quantity 500mL
MDL Number MFCD00000867
UN Number UN2356
Beilstein 1730782
Merck Index 14,5211
Synonym isopropyl chloride, propane, 2-chloro, isopropylchloride, 2-propyl chloride, sec-propyl chloride, chlorodimethylmethane, narcosop, isoprid, unii-vu39j8aj2n, ccris 874
InChI Key ULYZAYCEDJDHCC-UHFFFAOYSA-N
SMILES CC(C)Cl
IUPAC Name 2-chloropropane
Molecular Weight (g/mol) 78.539
PubChem CID 6361
Formula Weight 78.54
Percent Purity 99%
Chemical Name or Material 2-Chloropropane
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H224-H301-H311-H332
Extremely flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
P210-P301+P310-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: 2-CHLOROPROPANE
RTECSNumber TX4410000
TSCA TSCA
Recommended Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.