CAS RN 75-50-3
Trimethylamine, pure, 7.3M (50 wt.%) aqueous solution
CAS: 75-50-3 | C3H9N | 59.11 g/mol
Trimethylamine, pure, 4.2M (33 wt.%) solution in ethanol
CAS: 75-50-3 | C3H9N | 59.11 g/mol
Trimethylamine, 33% w/w in ethanol denatured with 2% cyclohexane
CAS: 75-50-3 Molecular Formula: C3H9N Molecular Weight (g/mol): 59.11 MDL Number: MFCD00008327 InChI Key: GETQZCLCWQTVFV-UHFFFAOYSA-N Synonym: trimethylamine,methanamine, n,n-dimethyl,dimethylmethaneamine,n-trimethylamine,trimethylamine solution,ch3 3n,trimethyl amine,trimethylamin,trimethyl-amine,fema number 3241 PubChem CID: 1146 ChEBI: CHEBI:18139 IUPAC Name: N,N-dimethylmethanamine SMILES: CN(C)C
Trimethylamine, 1M soln. in THF
CAS: 75-50-3 Molecular Formula: C3H9N Molecular Weight (g/mol): 59.11 MDL Number: MFCD00008327 InChI Key: GETQZCLCWQTVFV-UHFFFAOYSA-N Synonym: trimethylamine,methanamine, n,n-dimethyl,dimethylmethaneamine,n-trimethylamine,trimethylamine solution,ch3 3n,trimethyl amine,trimethylamin,trimethyl-amine,fema number 3241 PubChem CID: 1146 ChEBI: CHEBI:18139 IUPAC Name: N,N-dimethylmethanamine SMILES: CN(C)C
Trimethylamine, 1M solution in THF, AcroSeal™
CAS: 75-50-3 | C3H9N | 59.11 g/mol
Trimethylamine, 45% w/w aq. soln.
CAS: 75-50-3 Molecular Formula: C3H9N Molecular Weight (g/mol): 59.11 MDL Number: MFCD00008327 InChI Key: GETQZCLCWQTVFV-UHFFFAOYSA-N Synonym: trimethylamine,methanamine, n,n-dimethyl,dimethylmethaneamine,n-trimethylamine,trimethylamine solution,ch3 3n,trimethyl amine,trimethylamin,trimethyl-amine,fema number 3241 PubChem CID: 1146 ChEBI: CHEBI:18139 SMILES: CN(C)C
Trimethylamine (ca. 25% in Methanol, ca. 3.2mol/L), TCI America™
CAS: 75-50-3 Molecular Formula: C3H9N Molecular Weight (g/mol): 59.11 MDL Number: MFCD00008327 InChI Key: GETQZCLCWQTVFV-UHFFFAOYSA-N Synonym: trimethylamine,methanamine, n,n-dimethyl,dimethylmethaneamine,n-trimethylamine,trimethylamine solution,ch3 3n,trimethyl amine,trimethylamin,trimethyl-amine,fema number 3241 PubChem CID: 1146 ChEBI: CHEBI:18139 IUPAC Name: trimethylamine SMILES: CN(C)C
Trimethylamine (ca. 28% in Water, ca. 4.3mol/L), TCI America™
CAS: 75-50-3 Molecular Formula: C3H9N Molecular Weight (g/mol): 59.11 MDL Number: MFCD00008327 InChI Key: GETQZCLCWQTVFV-UHFFFAOYSA-N Synonym: trimethylamine,methanamine, n,n-dimethyl,dimethylmethaneamine,n-trimethylamine,trimethylamine solution,ch3 3n,trimethyl amine,trimethylamin,trimethyl-amine,fema number 3241 PubChem CID: 1146 ChEBI: CHEBI:18139 IUPAC Name: trimethylamine SMILES: CN(C)C
Trimethylamine (ca. 25% in Isopropyl Alcohol, ca. 3mol/L), TCI America™
CAS: 75-50-3 Molecular Formula: C3H9N Molecular Weight (g/mol): 59.11 MDL Number: MFCD00008327 InChI Key: GETQZCLCWQTVFV-UHFFFAOYSA-N Synonym: trimethylamine,methanamine, n,n-dimethyl,dimethylmethaneamine,n-trimethylamine,trimethylamine solution,ch3 3n,trimethyl amine,trimethylamin,trimethyl-amine,fema number 3241 PubChem CID: 1146 ChEBI: CHEBI:18139 IUPAC Name: trimethylamine SMILES: CN(C)C
Trimethylamine (ca. 25% in Ethanol, ca. 3mol/L), TCI America™
CAS: 75-50-3 Molecular Formula: C3H9N Molecular Weight (g/mol): 59.11 MDL Number: MFCD00008327 InChI Key: GETQZCLCWQTVFV-UHFFFAOYSA-N Synonym: trimethylamine,methanamine, n,n-dimethyl,dimethylmethaneamine,n-trimethylamine,trimethylamine solution,ch3 3n,trimethyl amine,trimethylamin,trimethyl-amine,fema number 3241 PubChem CID: 1146 ChEBI: CHEBI:18139 IUPAC Name: trimethylamine SMILES: CN(C)C
Trimethylamine (ca. 13% in Tetrahydrofuran, ca. 2mol/L), TCI America™
CAS: 75-50-3 Molecular Formula: C3H9N Molecular Weight (g/mol): 59.11 MDL Number: MFCD00008327 InChI Key: GETQZCLCWQTVFV-UHFFFAOYSA-N Synonym: trimethylamine,methanamine, n,n-dimethyl,dimethylmethaneamine,n-trimethylamine,trimethylamine solution,ch3 3n,trimethyl amine,trimethylamin,trimethyl-amine,fema number 3241 PubChem CID: 1146 ChEBI: CHEBI:18139 IUPAC Name: trimethylamine SMILES: CN(C)C
Trimethylamine, SPEX CertiPrep™
CAS: 75-50-3 Molecular Formula: C3H9N Molecular Weight (g/mol): 59.11 MDL Number: MFCD00008327 InChI Key: GETQZCLCWQTVFV-UHFFFAOYSA-N PubChem CID: 1146 ChEBI: CHEBI:18139 IUPAC Name: N,N-dimethylmethanamine SMILES: CN(C)C
Sigma Aldrich Trimethylamine solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Trimethylamine solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Trimethylamine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
- 1
- 2