Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylamine (ca. 25% in Ethanol, ca. 3mol/L), TCI America™
SDP

Catalog No. T2892100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2892100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T2892100ML Supplier TCI America Supplier No. T2892100ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An ethanol solution and is supplied in a bottle equipped with a Dual Seal septum-type cap.

Chemical Identifiers

CAS 75-50-3
Molecular Formula C3H9N
Molecular Weight (g/mol) 59.11
MDL Number MFCD00008327
InChI Key GETQZCLCWQTVFV-UHFFFAOYSA-N
Synonym trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241
PubChem CID 1146
ChEBI CHEBI:18139
IUPAC Name trimethylamine
SMILES CN(C)C

Specifications

Color Colorless
Quantity 100 mL
UN Number 2924
Formula Weight 59.11
Grade Not Available
Physical Form Liquid
Chemical Name or Material Trimethylamine (ca. 25% in Ethanol, ca. 3mol/L)
EINECSNumber (2)-0140
RTECSNumber PA0350000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.