CAS RN 75-52-5
Nitromethane, 99+%, for analysis
CAS: 75-52-5 Molecular Formula: CH3NO2 Molecular Weight (g/mol): 61.04 MDL Number: MFCD00007400 InChI Key: LYGJENNIWJXYER-UHFFFAOYSA-N Synonym: methane, nitro,nitrocarbol,nitrometan,nitrometan polish,nitro-methane,unii-ru5wg8c3f4,ccris 1205,hsdb 106,meno2,ru5wg8c3f4 PubChem CID: 6375 ChEBI: CHEBI:77701 IUPAC Name: nitromethane SMILES: C[N+]([O-])=O
Nitromethane, 96%, for HPLC
CAS: 75-52-5 Molecular Formula: CH3NO2 Molecular Weight (g/mol): 61.04 MDL Number: MFCD00007400 InChI Key: LYGJENNIWJXYER-UHFFFAOYSA-N Synonym: methane, nitro,nitrocarbol,nitrometan,nitrometan polish,nitro-methane,unii-ru5wg8c3f4,ccris 1205,hsdb 106,meno2,ru5wg8c3f4 PubChem CID: 6375 ChEBI: CHEBI:77701 IUPAC Name: nitromethane SMILES: C[N+]([O-])=O
Nitromethane 98.0+%, TCI America™
CAS: 75-52-5 Molecular Formula: CH3NO2 Molecular Weight (g/mol): 61.04 MDL Number: MFCD00007400 InChI Key: LYGJENNIWJXYER-UHFFFAOYSA-N Synonym: methane, nitro,nitrocarbol,nitrometan,nitrometan polish,nitro-methane,unii-ru5wg8c3f4,ccris 1205,hsdb 106,meno2,ru5wg8c3f4 PubChem CID: 6375 ChEBI: CHEBI:77701 IUPAC Name: nitromethane SMILES: C[N+]([O-])=O
Nitromethane, 96%, ACS reagent
CAS: 75-52-5 Molecular Formula: CH3NO2 Molecular Weight (g/mol): 61.04 MDL Number: MFCD00007400 InChI Key: LYGJENNIWJXYER-UHFFFAOYSA-N Synonym: methane, nitro,nitrocarbol,nitrometan,nitrometan polish,nitro-methane,unii-ru5wg8c3f4,ccris 1205,hsdb 106,meno2,ru5wg8c3f4 PubChem CID: 6375 ChEBI: CHEBI:77701 IUPAC Name: nitromethane SMILES: C[N+]([O-])=O
Nitromethane, 98+%
CAS: 75-52-5 Molecular Formula: CH3NO2 Molecular Weight (g/mol): 61.04 MDL Number: MFCD00007400 InChI Key: LYGJENNIWJXYER-UHFFFAOYSA-N Synonym: methane, nitro,nitrocarbol,nitrometan,nitrometan polish,nitro-methane,unii-ru5wg8c3f4,ccris 1205,hsdb 106,meno2,ru5wg8c3f4 PubChem CID: 6375 ChEBI: CHEBI:77701 IUPAC Name: nitromethane SMILES: C[N+]([O-])=O
Nitromethane, 96%, for spectroscopy
CAS: 75-52-5 Molecular Formula: CH3NO2 Molecular Weight (g/mol): 61.04 MDL Number: MFCD00007400 InChI Key: LYGJENNIWJXYER-UHFFFAOYSA-N Synonym: methane, nitro,nitrocarbol,nitrometan,nitrometan polish,nitro-methane,unii-ru5wg8c3f4,ccris 1205,hsdb 106,meno2,ru5wg8c3f4 PubChem CID: 6375 ChEBI: CHEBI:77701 IUPAC Name: nitromethane SMILES: C[N+]([O-])=O
Nitromethane, 99%, pure
CAS: 75-52-5 Molecular Formula: CH3NO2 Molecular Weight (g/mol): 61.04 MDL Number: MFCD00007400 InChI Key: LYGJENNIWJXYER-UHFFFAOYSA-N Synonym: methane, nitro,nitrocarbol,nitrometan,nitrometan polish,nitro-methane,unii-ru5wg8c3f4,ccris 1205,hsdb 106,meno2,ru5wg8c3f4 PubChem CID: 6375 ChEBI: CHEBI:77701 IUPAC Name: nitromethane SMILES: C[N+]([O-])=O
Nitromethane, ACS, 95%, Spectrum™ Chemical
CAS: 75-52-5 Molecular Formula: CH3NO2 Molecular Weight (g/mol): 61.04 InChI Key: LYGJENNIWJXYER-UHFFFAOYSA-N IUPAC Name: nitromethane SMILES: C[N+]([O-])=O
Nitromethane (55% in Methanol, ca. 8.7mol/L), TCI America™
CAS: 75-52-5 Molecular Formula: CH3NO2 Molecular Weight (g/mol): 61.04 MDL Number: MFCD00007400 InChI Key: LYGJENNIWJXYER-UHFFFAOYSA-N Synonym: methane, nitro,nitrocarbol,nitrometan,nitrometan polish,nitro-methane,unii-ru5wg8c3f4,ccris 1205,hsdb 106,meno2,ru5wg8c3f4 PubChem CID: 6375 ChEBI: CHEBI:77701 IUPAC Name: nitromethane SMILES: C[N+]([O-])=O
Nitromethane 98.0+%, TCI America™
CAS: 75-52-5 Molecular Formula: CH3NO2 Molecular Weight (g/mol): 61.04 MDL Number: MFCD00007400 InChI Key: LYGJENNIWJXYER-UHFFFAOYSA-N Synonym: methane, nitro,nitrocarbol,nitrometan,nitrometan polish,nitro-methane,unii-ru5wg8c3f4,ccris 1205,hsdb 106,meno2,ru5wg8c3f4 PubChem CID: 6375 ChEBI: CHEBI:77701 IUPAC Name: nitromethane SMILES: C[N+]([O-])=O
Nitromethane, ≥99% (GC), MilliporeSigma™ Supelco™
Nitromethane belongs to the class of residual solvents which are volatile organic compounds (VOCs) that are employed in manufacturing processes of pharmaceutical industries.
Nitromathan in Dimethyl Sulfoxide, SPEX CertiPrep™
CAS: 75-52-5 Molecular Formula: CH3NO2 Molecular Weight (g/mol): 61.04 MDL Number: MFCD00007400 InChI Key: LYGJENNIWJXYER-UHFFFAOYSA-N PubChem CID: 6375 ChEBI: CHEBI:77701 IUPAC Name: nitromethane SMILES: C[N+]([O-])=O