Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitromethane, ACS, 95%, Spectrum™ Chemical
SDP

Catalog No. 18612029 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
55 gal.
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-029 4 L Amber Glass Bottle
18-612-026 500 mL Amber Glass Bottle
18-612-028 20 L Amber Glass Bottle
18-612-027 55 gal. Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-612-029 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. N10954LTGL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 75-52-5
Molecular Formula CH3NO2
Molecular Weight (g/mol) 61.04
InChI Key LYGJENNIWJXYER-UHFFFAOYSA-N
IUPAC Name nitromethane
SMILES C[N+]([O-])=O

Specifications

CAS Max % 1
Boiling Point 101.1°C (214°F)
Packaging Amber Glass Bottle
Quantity 4 L
Percent Purity 0.95
Grade ACS
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material nitromethane
WARNING: Cancer - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.