accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFBrBr

CAS RN 75-61-6

IUPAC Name: dibromodifluoromethane
Synonyms: dibromodifluoromethane
Molecular Weight (g/mol): 209.82
Molecular Formula: CBr2F2
InChi Key: AZSZCFSOHXEJQE-UHFFFAOYSA-N
SMILES: FC(F)(Br)Br
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)