CAS RN 75-65-0
tert-Butanol, 99.5%, extra pure
CAS: 75-65-0 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.123 MDL Number: MFCD00004464 InChI Key: DKGAVHZHDRPRBM-UHFFFAOYSA-N Synonym: tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol PubChem CID: 6386 ChEBI: CHEBI:45895 IUPAC Name: 2-methylpropan-2-ol SMILES: CC(C)(C)O
tert-Butyl alcohol, 99%
CAS: 75-65-0 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.123 MDL Number: MFCD00004464 InChI Key: DKGAVHZHDRPRBM-UHFFFAOYSA-N Synonym: tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol PubChem CID: 6386 ChEBI: CHEBI:45895 IUPAC Name: 2-methylpropan-2-ol SMILES: CC(C)(C)O
tert-Butyl alcohol, ACS, 99+%
CAS: 75-65-0 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.123 MDL Number: MFCD00004464 InChI Key: DKGAVHZHDRPRBM-UHFFFAOYSA-N Synonym: tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol PubChem CID: 6386 ChEBI: CHEBI:45895 IUPAC Name: 2-methylpropan-2-ol SMILES: CC(C)(C)O
tert-Butanol (Certified), Fisher Chemical™
CAS: 75-65-0 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.123 MDL Number: MFCD00004464 InChI Key: DKGAVHZHDRPRBM-UHFFFAOYSA-N Synonym: tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol PubChem CID: 6386 ChEBI: CHEBI:45895 IUPAC Name: 2-methylpropan-2-ol SMILES: CC(C)(C)O
tert-Butyl alcohol, anhydrous, 99.5%
CAS: 75-65-0 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.123 MDL Number: MFCD00004464 InChI Key: DKGAVHZHDRPRBM-UHFFFAOYSA-N Synonym: tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol PubChem CID: 6386 ChEBI: CHEBI:45895 IUPAC Name: 2-methylpropan-2-ol SMILES: CC(C)(C)O
tert-Butyl alcohol, anhydrous, 99.5%, packaged under Argon in resealable ChemSeal™ bottles
CAS: 75-65-0 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.123 MDL Number: MFCD00004464 InChI Key: DKGAVHZHDRPRBM-UHFFFAOYSA-N Synonym: tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol PubChem CID: 6386 ChEBI: CHEBI:45895 IUPAC Name: 2-methylpropan-2-ol SMILES: CC(C)(C)O
tert-Butanol, ACS reagent
CAS: 75-65-0 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.123 MDL Number: MFCD00004464 InChI Key: DKGAVHZHDRPRBM-UHFFFAOYSA-N Synonym: tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol PubChem CID: 6386 ChEBI: CHEBI:45895 IUPAC Name: 2-methylpropan-2-ol SMILES: CC(C)(C)O
tert-Butanol, 99.5%, for analysis
CAS: 75-65-0 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.123 InChI Key: DKGAVHZHDRPRBM-UHFFFAOYSA-N Synonym: tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol PubChem CID: 6386 ChEBI: CHEBI:45895 IUPAC Name: 2-methylpropan-2-ol SMILES: CC(C)(C)O
tert-Butanol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards
tert-Butyl Alcohol ACS AR, Macron Fine Chemicals™
CAS: 75-65-0 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.123 InChI Key: DKGAVHZHDRPRBM-UHFFFAOYSA-N Synonym: tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol PubChem CID: 6386 ChEBI: CHEBI:45895 IUPAC Name: 2-methylpropan-2-ol SMILES: CC(C)(C)O
tert-Butanol, Analytical Standard, ≥99.8% (GC), MilliporeSigma™ Supelco™
tert-butanol can find applications as an ideal reaction medium or solvent for the lipase-catalyzed biodiesel production.
tert-Butyl Alcohol, ACS, 99.0%, MilliporeSigma™
CAS: 75-65-0 Molecular Weight (g/mol): 74.12
tert-Butyl Alcohol, Reagent, 99%, Spectrum™ Chemical
CAS: 75-65-0 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.12 InChI Key: DKGAVHZHDRPRBM-UHFFFAOYSA-N IUPAC Name: 2-methylpropan-2-ol SMILES: CC(C)(C)O
tert-Butyl Alcohol, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 75-65-0 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.123 InChI Key: DKGAVHZHDRPRBM-UHFFFAOYSA-N Synonym: tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol PubChem CID: 6386 ChEBI: CHEBI:45895 IUPAC Name: 2-methylpropan-2-ol SMILES: CC(C)(C)O
Tert-Butanol, Certified Reference Material, MilliporeSigma™ Supelco™
These secondary standards offer multi-traceability to the USP, EP, and BP Primary Standards, where they are available.
- 1
- 2