Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tert-Butanol, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023040 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-3040 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-3040 Supplier MilliporeSigma Supelco Supplier No. RTC00008220ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These secondary standards offer multi-traceability to the USP, EP, and BP Primary Standards, where they are available.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 23°C to 26°C (literature)
CAS 75-65-0
Density 0.775 g/mL (at 25°C (literature))
Quantity 20 mL
Flash Point 15°C (Closed Cup)
Refractive Index n20/D 1.387 (literature)
Molecular Formula C4H10O
Linear Formula (CH3)3 COH
Vapor Pressure 31 mmHg (20°C)
MDL Number MFCD00004464
Synonym tert-Butanol; 2-Methyl-2-propanol; tert-Butyl alcohol; Trimethyl carbinol
Boiling Point 83°C (literature)
Formula Weight 74.12
Grade Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.