accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 75-71-8

FFClCl

CAS RN 75-71-8

IUPAC Name: dichlorodifluoromethane
Synonyms: dichlorodifluoromethane
Molecular Weight (g/mol): 120.91
Molecular Formula: CCl2F2
InChi Key: PXBRQCKWGAHEHS-UHFFFAOYSA-N
SMILES: FC(F)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results