CAS RN 75-76-3
Tetramethylsilane, 99.9+%, NMR grade
CAS: 75-76-3 Molecular Formula: C4H12Si Molecular Weight (g/mol): 88.23 MDL Number: MFCD00008274 InChI Key: CZDYPVPMEAXLPK-UHFFFAOYSA-N Synonym: silane, tetramethyl,tetramethylsilicane,tetramethyl silane,silicon, tetramethyl,tetramethyl-silane,unii-41y0rbg14q,me4si,ch3 4si,si ch3 4,chembl68073 PubChem CID: 6396 ChEBI: CHEBI:85361 IUPAC Name: tetramethylsilane SMILES: C[Si](C)(C)C
Tetramethylsilane, 99%
CAS: 75-76-3 Molecular Formula: C4H12Si Molecular Weight (g/mol): 88.23 MDL Number: MFCD00008274 InChI Key: CZDYPVPMEAXLPK-UHFFFAOYSA-N Synonym: silane, tetramethyl,tetramethylsilicane,tetramethyl silane,silicon, tetramethyl,tetramethyl-silane,unii-41y0rbg14q,me4si,ch3 4si,si ch3 4,chembl68073 PubChem CID: 6396 ChEBI: CHEBI:85361 IUPAC Name: tetramethylsilane SMILES: C[Si](C)(C)C
Tetramethylsilane, 99%, AcroSeal™
CAS: 75-76-3 Molecular Formula: C4H12Si Molecular Weight (g/mol): 88.23 MDL Number: MFCD00008274 InChI Key: CZDYPVPMEAXLPK-UHFFFAOYSA-N Synonym: silane, tetramethyl,tetramethylsilicane,tetramethyl silane,silicon, tetramethyl,tetramethyl-silane,unii-41y0rbg14q,me4si,ch3 4si,si ch3 4,chembl68073 PubChem CID: 6396 ChEBI: CHEBI:85361 IUPAC Name: tetramethylsilane SMILES: C[Si](C)(C)C
Tetramethylsilane, for the calibration of NMR spectra for NMR spectroscopy, MagniSolv™, MilliporeSigma™
CAS: 75-76-3 Molecular Formula: C4H12Si Molecular Weight (g/mol): 88.23 MDL Number: MFCD00008274 InChI Key: CZDYPVPMEAXLPK-UHFFFAOYSA-N Synonym: silane, tetramethyl,tetramethylsilicane,tetramethyl silane,silicon, tetramethyl,tetramethyl-silane,unii-41y0rbg14q,me4si,ch3 4si,si ch3 4,chembl68073 PubChem CID: 6396 ChEBI: CHEBI:85361 IUPAC Name: tetramethylsilane SMILES: C[Si](C)(C)C
Tetramethylsilane, 99.9%
CAS: 75-76-3 Molecular Formula: C4H12Si Molecular Weight (g/mol): 88.23 MDL Number: MFCD00008274 InChI Key: CZDYPVPMEAXLPK-UHFFFAOYSA-N Synonym: silane, tetramethyl,tetramethylsilicane,tetramethyl silane,silicon, tetramethyl,tetramethyl-silane,unii-41y0rbg14q,me4si,ch3 4si,si ch3 4,chembl68073 PubChem CID: 6396 ChEBI: CHEBI:85361 IUPAC Name: tetramethylsilane SMILES: C[Si](C)(C)C
Tetramethylsilane, ACS reagent, NMR grade
CAS: 75-76-3 Molecular Formula: C4H12Si Molecular Weight (g/mol): 88.23 MDL Number: MFCD00008274 InChI Key: CZDYPVPMEAXLPK-UHFFFAOYSA-N Synonym: silane, tetramethyl,tetramethylsilicane,tetramethyl silane,silicon, tetramethyl,tetramethyl-silane,unii-41y0rbg14q,me4si,ch3 4si,si ch3 4,chembl68073 PubChem CID: 6396 ChEBI: CHEBI:85361 IUPAC Name: tetramethylsilane SMILES: C[Si](C)(C)C
Tetramethylsilane 99.0+%, TCI America™
CAS: 75-76-3 Molecular Formula: C4H12Si Molecular Weight (g/mol): 88.23 MDL Number: MFCD00008274 InChI Key: CZDYPVPMEAXLPK-UHFFFAOYSA-N Synonym: silane, tetramethyl,tetramethylsilicane,tetramethyl silane,silicon, tetramethyl,tetramethyl-silane,unii-41y0rbg14q,me4si,ch3 4si,si ch3 4,chembl68073 PubChem CID: 6396 ChEBI: CHEBI:85361 IUPAC Name: tetramethylsilane SMILES: C[Si](C)(C)C
Sigma Aldrich Tetramethylsilane
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Tetramethylsilane
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Tetramethylammonium Hydrogen Sulfate Monohydrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich N,N,N',N'-Tetramethylethylenediamine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.