accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFCl

CAS RN 75-88-7

IUPAC Name: 2-chloro-1,1,1-trifluoroethane
Synonyms: 1,1,1-trifluoro-2-chloroethane
Molecular Weight (g/mol): 118.48
Molecular Formula: C2H2ClF3
InChi Key: CYXIKYKBLDZZNW-UHFFFAOYSA-N
SMILES: FC(F)(F)CCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results