accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHNH2I

CAS RN 75073-11-9

IUPAC Name: 6-amino-3-iodo-2-methylpyridin-1-ium
Synonyms: 6-amino-3-iodo-2-methylpyridin-1-ium
Molecular Weight (g/mol): 235.05
Molecular Formula: C6H8IN2
InChi Key: AFMNJDHAAFAELF-UHFFFAOYSA-O
SMILES: CC1=[NH+]C(N)=CC=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results