accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7508-82-9

CH3(R)HOHNOOO

CAS RN 7508-82-9

IUPAC Name: (2R)-3-hydroxy-2-methyl-2-(phenylformamido)propanoate
Synonyms: (2R)-3-hydroxy-2-methyl-2-(phenylformamido)propanoate
Molecular Weight (g/mol): 222.22
Molecular Formula: C11H12NO4
InChi Key: FVWQRKJTTRAXJP-LLVKDONJSA-M
SMILES: C[C@](CO)(NC(=O)C1=CC=CC=C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results