accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 75148-49-1

IUPAC Name: 1-bromo-3-(diethoxymethyl)benzene
Synonyms: 1-bromo-3-(diethoxymethyl)benzene
Molecular Weight (g/mol): 259.14
Molecular Formula: C11H15BrO2
InChi Key: KDHRJLQXMOBXRV-UHFFFAOYSA-N
SMILES: CCOC(OCC)C1=CC=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results