Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromobenzaldehyde Diethyl Acetal 98.0+%, TCI America™
SDP

Catalog No. B382225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B382225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B382225G Supplier TCI America Supplier No. B382225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 75148-49-1
Molecular Formula C11H15BrO2
Molecular Weight (g/mol) 259.14
MDL Number MFCD01075697
InChI Key KDHRJLQXMOBXRV-UHFFFAOYSA-N
Synonym 3-bromobenzaldehyde diethyl acetal, 1-bromo-3-diethoxymethyl benzene, 3-bromophenyl diethoxymethane, benzene, 1-bromo-3-diethoxymethyl, 3-bromo-alpha,alpha-diethoxytoluene, 3-bromobenzaldehydediethylacetal, pubchem5371, acmc-209ox6, 1-diethoxymethyl-3-bromobenzene, 1-bromo-3-diethoxymethyl-benzene
PubChem CID 2733543
IUPAC Name 1-bromo-3-(diethoxymethyl)benzene
SMILES CCOC(OCC)C1=CC=CC(Br)=C1

Specifications

Color Yellow
Boiling Point 95°C
Quantity 25 g
Formula Weight 259.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromobenzaldehyde Diethyl Acetal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.