accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOOOOOONHN(R)H3CCH3CH3

CAS RN 75179-29-2

IUPAC Name: 2,5-dioxopyrrolidin-1-yl (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-[(triphenylmethyl)sulfanyl]propanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl (2R)-2-[(tert-butoxycarbonyl)amino]-3-[(triphenylmethyl)sulfanyl]propanoate
Molecular Weight (g/mol): 560.67
Molecular Formula: C31H32N2O6S
InChi Key: JIXAKJALBJDZTP-VWLOTQADSA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)